Adapter, Twin Hose Connection, Ace Glass Incorporated
Supplier: Ace Glass
5206-04 5206-12 5206-20 5206-10 5206-114 5206-124
89082-804EA 95.24 CAD
89082-804 89082-806 89082-808 89051-412 10098-740 10098-742
Adapter, Twin Hose Connection, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This adapter is normally used with a reflux apparatus to allow inert gas flow over top of the apparatus to maintain an oxygen free system.
  • [ST] inner joint
  • Twin hose connections opposite to each other
  • Fabricated from borosilicate glass
This adapter has a standard taper inner joint and twin hose connections opposite to each other. Use with a 3/8" or 5/16" inner diameter tubing, size C hose connection.