Spherical Jacketed Reaction Flask, Ace Glass Incorporated
Supplier: Ace Glass
6536-06 6536-09 6536-11 6536-309 6536-311 6536-306 6536-55
89055-478EA 3848.94 CAD
89055-478 89055-480 89055-482 10098-974 10098-976 10099-402 10767-918
Spherical Jacketed Reaction Flask, Ace Glass Incorporated
Flasks Round Bottom Flasks
This rugged, spherical reaction flask with Duran style flat flange has an O-ring groove for use with the Ace Glass quick release clamp
The 0-14 Easy-Action and Piston bottoms have a 28/15 ball joint bottom connector. Each flask is supplied with one CAPFE (PTFE encapsulated silicone) O-ring. Plain silicone O-rings are also available. All capacities have size 28/15 O-ring ball joints on inlet and outlet, sealed tangentially. Fabricated from borosilicate glass.