Reaction Head, Duran® Flange, Ace Glass Incorporated
Supplier: Ace Glass
6529-32 6529-36 6529-39 6529-23 6529-25 6529-27 6529-29 6529-03 6529-04 6529-05 6529-06
89055-326EA 1352.22 CAD
89055-326 89055-328 89055-330 89084-080 89084-082 89084-084 89084-086 89084-972 89084-976 89084-980 89084-984
Reaction Head, Duran® Flange, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
This flat ground Duran® style flange can be used with all reaction flasks that have a 150 mm Duran® ground mating flange
Supplied with four ground joints. Fabricated from borosilicate glass.
Ordering information: For clamp, order #89055-112 .