Conical Flange Reaction Head, Ace Glass
Supplier: Ace Glass
6486-10 6486-20 6486-25 6486-40
89054-962EA 606.75 CAD
89054-962 89054-964 89054-966 89054-968
Conical Flange Reaction Head, Ace Glass
Reaction Vessels Reaction Vessels, Flask Shape
Standard 4" (100 mm), three-outlet head for use with cetain ACE reaction flasks
Supplied with three standard taper outer joints. Fabricated from borosilicate glass.