Jacketed Reaction Flasks, Ace Glass
Supplier: Ace Glass
6519-01 6519-02 6519-03 6519-04 6519-301 6519-302 6519-303 6519-304
89215-006EA 2089.14 CAD
89215-006 89215-016 89215-020 89215-024 10098-966 10098-968 10098-970 10098-972
Jacketed Reaction Flasks, Ace Glass
Reaction Vessels Reaction Vessels, Flask Shape
This is a cylindrical, heavy-wall reaction flask, except flat flange has an O-ring groove for use with CAPFE® (PTFE encapsulated silicone rubber) O-ring instead of a gasket
Without constriction at top to facilitate introduction/removal of material and allow for ease of cleaning. Flat bottom rounded into side wall. Flask is supplied with one CAPFE O-ring. Comes with 28/15 O‑ring ball joint for easy connect/disconnect from hoses, or #11 Ace-Thred. Made from borosilicate glass.