SP Bel-Art 72-Piece Complete Fitting Assortment, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F19625-0000 F19625-0000_SE
62866-170EA 441.8 CAD
62866-170 470123-372
SP Bel-Art 72-Piece Complete Fitting Assortment, Bel-Art Products, a part of SP
Fittings and Connectors
This generous assortment of 72 plastic fittings has everything you need for routine laboratory set-ups.
  • Clear Polystyrene Construction
  • Hinged Lid
  • Translucent Siding
  • Seventy-Two Plastic Fittings Included
  • Autoclavable Fittings
A clear, hinge-top, polystyrene storage box makes it easy to find what you need when you need it.
The kit includes common sizes of connector and stopper styles.
Certifications: Polypropylene fittings are FDA-graded for direct contact with food, drugs, and cosmetics and are autoclavable.
Ordering information: ThisMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable