Orion™ Conductivity Standards, Solutions, and Accessories, Thermo Scientific
Supplier: Thermo Fisher Scientific
990106 011005 011007 011006 011005
23227-504EA 186.9 CAD
23227-504 CA011005 52458-147 52458-145 71002-397
Orion™ Conductivity Standards, Solutions, and Accessories, Thermo Scientific
Standards Conductivity Standards
Perform virtually any electrochemistry analysis with Orion™ Conductivity Standards and Solutions
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
Conductivity standard temperature tables are programmed into Orion™ meter software for use with the automatic calibration feature.
Choose from a complete selection of conductivity accessories including conductivity standards and TDS (total dissolved solids) standards. All conductivity standards are traceable to NIST standard reference materials.