Whatman™ Kjeldahl Analysis Weighing Paper, Whatman products (Cytiva)
Supplier: Cytiva
10347670 10347671 10347672 10347673 10347670 10347671 10347672 10347673
28498-000PK 339.66 CAD
28498-000 28498-002 28498-004 28498-006 CA28498-000 CA28498-002 CA28498-004 CA28498-006
Whatman™ Kjeldahl Analysis Weighing Paper, Whatman products (Cytiva)
Weighing Papers
Grade B-2 Whatman weighing paper from Cytiva dissolves residue-free in acid solution, which is critical in applications such as Kjeldahl analysis of nitrogen in soil.
  • Thin (0.04 mm) and lightweight (43 g/m²)
  • Transparent and smooth parchment surface
  • Simple weighing out and sample transfer
  • Reliable for quantitative transfer
  • Range of sheet sizes for ease-of-use
  • Low nitrogen paper that is residue-free for accurate chemical analysis
Cytiva also offers weighing boats for Kjeldahl analysis.
Packs of 500 Grade B-2 Whatman weighing paper are available in four sheet sizes: 76.2×76.2 mm (3×3 inch) sheets; 101.6×101.6 mm (4×4 inch) sheets; 152.4×152.4 mm (6×6 inch) sheets; 304.8×304.8 mm (12×12 inch) sheets.