Simmons Citrate Agars, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES PVT. CA
M099-500G M411-100G M411-500G M099-5KG M099-2.5KG M099S-2.5KG M099S-5KG
CA61000-012EA 868.61 CAD
CA61000-012 CA61000-104 CA95021-790 CA95026-812 CA89222-552 CA89222-554 CA89222-556
Simmons Citrate Agars, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Simmons Citrate Agar is for the differentiation between microbiology members on the basis of citrate utilization.