Trubore® Bearing Ace-Thred Turbing, Ace Glass Incorporated
Supplier: Ace Glass
8043-08 8043-16 8043-20 8043-30 8043-45
89061-780EA 293.27 CAD
89061-780 89061-782 89061-784 89061-786 89061-788
Trubore® Bearing Ace-Thred Turbing, Ace Glass Incorporated
Stirrers Stirring Tools
Designed for use with #25 Ace-Thred bushing, precision ground stirring shafts
Delivery information: Consists of a Trubore® bearing, pressure bushing, gland with two FETFE o-rings and retainer bushing with FETFE o-ring.