PTFE Bearing/Debris Trap, Ace Glass Incorporated
Supplier: Ace Glass
8067-45 8067-47 8067-49
89084-806EA 521.85 CAD
89084-806 89084-808 89084-810
PTFE Bearing/Debris Trap, Ace Glass Incorporated
Stirrers Stirring Shafts
Combination all-in-one stirring shaft bearing and debris trap
Designed to prevent particles from entering the reaction vessel. The debris trap section consists of a PTFE washer and particle well. #11 Ace-Thred on glass body side allows access for clean-out, or may be used for evacuation/gas purge port. Complete item supplied with glass body, inner PTFE bearing, PTFE bushing and lock nut, Rulon saddle O-ring, “Ace-Safe” connector for 3/8" tubing, and PTFE plug.