Krytox GPL, Ace Glass Incorporated
Supplier: Ace Glass
8115-08
89062-220EA 234.59 CAD
89062-220
Krytox GPL, Ace Glass Incorporated
Lubricants
Superior performance, non-contaminating, nonflammable, general purpose grease
Krytox GPL grease has demonstrated an exceptional degree of inertness when contacted with a wide range of reactive chemicals. There is no reaction with the following chemicals: oxygen, caustic, fluorine, hydrazine, diethylene triamine, hydrocarbons, chlorine, hydrogen, ethanol, hydrogen peroxide, phosphoric acid, red fuming nitric acid, sulfuric acid, methanol, aniline, ammonia, hydrochloric acid, unsymmetrical dimethyl hydrazine
Krytox GPL can be used at operating temperatures up to 204°C (400°F) for extended periods of time and at 290°C (550°F) intermittently. Approximate minimum use temperature is -35°F.
Excellent as a glass bearing lubricant, as a super-inert grease for stopcocks and joints, as a high temperature grease in “baked-out” vacuum systems, or on distillation column joints because it is insoluble in almost all solvents except Freon® 113. Easy removal with fluorinated solvents.