304 Stainless Steel Beaded Pipe to Female NPT Adapter, Ace Glass Incorporated
Supplier: Ace Glass
8871-20 8871-22 8871-24 8871-28
89097-228EA 747.08 CAD
89097-228 89097-230 89097-232 89097-234
304 Stainless Steel Beaded Pipe to Female NPT Adapter, Ace Glass Incorporated
Fittings and Connectors
Adapter for matching beaded pipe to male NPT.
  • Beaded pipe to male NPT
  • 304 stainless steel
This is a 304 stainless steel adapter for matching glass beaded process pipe to male NPT fittings, gauges and flanges.
Order Now
SPECIFICATIONS
Environmentally Preferable