Graded Seals, Quartz to Borosilicate, Ace Glass Incorporated
Supplier: Ace Glass
8455-02 8455-04 8455-06 8455-08 8455-10 8455-12 8455-14 8455-16 8455-18 8455-20 8455-22 8455-24 8455-26 8455-28 8455-30 8455-32 8455-34 8455-36 8455-38 8455-40 8455-42 8455-44 8455-46 8455-48 8455-50
89063-046EA 169.12 CAD
89063-046 89063-048 89063-050 89063-052 89063-054 89063-056 89063-058 89063-060 89063-062 89063-064 89063-066 89063-068 89063-070 89063-072 89063-074 89063-076 89063-078 89063-080 89063-082 89063-084 89063-086 89063-088 89063-090 89063-092 89063-094
Graded Seals, Quartz to Borosilicate, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
For connecting Kimax, Pyrex, Simax or Duran 33 Expansion borosilicate glass parts to quartz
Quartz length is 7.6 cm, glass length, 10.2 cm.