Three-Piece Trubore® Bearing, Ace Glass Incorporated
Supplier: Ace Glass
8042-05 8042-09 8042-115 8042-117 8042-119 8042-121 8042-135 8042-19
89061-756EA 96.14 CAD
89061-756 89061-758 89061-760 89061-762 89061-764 89061-766 89061-768 89061-774
Three-Piece Trubore® Bearing, Ace Glass Incorporated
Stirrers Stirring Tools
This piece is a self-aligning three-piece bearing
  • Inner bearing and bushing can be used with any joint size
  • For use with 10mm outer diameter shafts
  • For replacement O-rings, order 89060-984
Complete consists of inner Trubore® glass bearing, nylon bushing with O-ring, and a threaded adapter. Unique design eliminates costly replacement since inner bearing will spin if shaft binds or will self-align at o-ring seal in the event that the motor is slightly cocked. Inner bearing and bushing can be used with any joint size. For use with 10mm outer diameterMore Product Information