Stirring Bearing, Vacuum, Ace Glass Incorporated
Supplier: Ace Glass
8099-03 8099-40 8099-38
89062-190EA 141.53 CAD
89062-190 89062-200 89062-206
Stirring Bearing, Vacuum, Ace Glass Incorporated
Stirrers Stirring Shafts
The upper member is internally threaded on the top
  • Easier to adjust shaft
  • suitable for vacuum work
  • For use with ACE stirring shafts
#7 Ace‑Thred nylon bushing and FETFE o-ring form a compression seal on stirring shaft in place of the conventional tubing arrangement.