(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) ≥99% (by GC), powder
Supplier: CHEM-IMPEX INTERNATIONAL, INC
Synonyms:
PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate
02276-100G
02276-1KG
02276-250G
02276-25G
CH02276-100GEA
116.46
CAD
CH02276-100G
CH02276-1KG
CH02276-250G
CH02276-25G
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) ≥99% (by GC), powder
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
One of the most convenient coupling reagents for peptide synthesis without carcinogenic by-products. It replaces BOP reagent. This is especially suitable for solid-phase peptide synthesis, avoids racemization.
Formula:
C₁₈H₂₈F₆N₆OP₂ MW: 520.4 g/mol |
MDL Number:
MFCD00077411 CAS Number: 128625-52-5 |
Specification Test Results
Molecular Formula | C18H28N6OP·F6P |
Molecular Weight | 520.34 |
Melting Point | 148-160 °C |
MDL Number | MFCD00077411 |
Storage Temperature | Store at RT |
Color | White to Off-White |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...