G418 disulfate

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: G418 disulfate, G-418 sulfate, G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate, G-418 disulfate

01417-1G 01417-5G
CH01417-1GEA 63.2 CAD
CH01417-1G CH01417-5G
G418 disulfate
G418 disulphate

This geneticin disulfate is plant cell culture, tested.


  • Potency: ≥ 700 mg/mg (anhydrous)


White to off-white powder

Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR