α-D-(+)-Melibiose ≥99%, powder

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: 6-O-alpha-D-Galactopyranosyl-D-glucose

00808-25G 00808-5G
CH00808-25GEA 122.64 CAD
CH00808-25G CH00808-5G
α-D-(+)-Melibiose ≥99%, powder
alpha-D-(+)-Melibiose
Formula: C₁₂H₂₂O₁₁
MW: 342.30 g/mol
MDL Number: MFCD00198188
CAS Number: 585-99-9

Order Now

Specification Test Results

Molecular Formula C12H22O11
Molecular Weight 342.3
Melting Point 182°C (dec)
Optical Rotation + 135 ± 2 ° (C = 4 in H2O)
MDL Number MFCD00198191
Storage Temperature Store at RT
Color White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR