Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt ≥99% (by UV method)

Supplier: CHEM-IMPEX INTERNATIONAL, INC

00616-100MG 00616-1G 00616-250MG 00616-5G
CH00616-100MGEA 252.8 CAD
CH00616-100MG CH00616-1G CH00616-250MG CH00616-5G
Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt ≥99% (by UV method)
Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt

  • Off-white lyophilized powder

Formula: C₂₁H₂₆N₇O₁₇P₃.4Na
MW: 833.348 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036263
CAS Number: 2646-71-1

Order Now

Specification Test Results

Molecular Formula C21H26N7O17P3Na4
Molecular Weight 833.4
MDL Number MFCD00036263
Storage Temperature Store at < 0 °C
Color Off-White


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR