ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form) ≥98%

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: Nadide phosphate disodium salt, Triphosphopyridine nucleotide, disodium salt, NADP-Na2, β-NADP-Na2, NADP disodium salt, Nicotinamide-adenine dinucleotide phosphate disodium salt, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium, disodium salt, β-NADH phosphate disodium salt

00513-1G 00513-5G
CH00513-1GEA 103.33 CAD
CH00513-1G CH00513-5G
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form) ≥98%
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)

ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)

Formula: C₂₁H₂₆N₇Na₂O₁₇P₃
MW: 787.37
Storage Temperature: Refrigerator
MDL Number: MFCD00065390
CAS Number: 24292-60-2

Order Now

Specification Test Results

Molecular Formula C21H26N7O17P3·Na2·2H2O
Molecular Weight 823.4
MDL Number MFCD00065390
Storage Temperature Store at ≤-4°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR