Amphotericin B (dried basis)

Supplier: CHEM-IMPEX INTERNATIONAL, INC

Synonyms: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid

00329-1G 00329-250MG 00329-25G
CH00329-1GEA 150.32 CAD
CH00329-1G CH00329-250MG CH00329-25G
Amphotericin B (dried basis)
Amphotericin B

Purity limit: ≥750 mg/mg (Microbial Assay on Dried Basis)


  • Yellow to orange powder

Formula: C₄₇H₇₃NO₁₇
MW: 924.08 g/mol
Melting Pt: >170 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00877763
CAS Number: 1397-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR