Rifampicin 97.0-103.0% (by HPLC)

Synonyms: Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP

00260-1G 00260-25G 00260-5G
CH00260-1GEA 31.6 CAD
CH00260-1G CH00260-25G CH00260-5G
Rifampicin 97.0-103.0% (by HPLC)
Rifampicin

This Rifampicin inhibits the assembly of D and protein into mature virus particles.


  • Brownish-red crystalline powder

Formula: C₄₃H₅₈N₄O₁₂
MW: 822.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00151389
CAS Number: 13292-46-1
Merck Index: 12,08382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR