Xylene Substitute, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
23410-05 23410-01 23410-04
100504-384EA 1222.16 CAD
100504-384 100504-382 100504-780
Xylene Substitute, Electron Microscopy Sciences
Stains Microscopy and staining reagents
This EMS Xylene Substitute solution is specially designed for use in processing and staining histological/cytological specimens. It is essentially odorless with low toxicity, less irritating to the skin than xylene- and d-limonene-based clearants. It can be substituted for xylene and can be used with almost all tissue processors and automated slide stainers.
  • Minimal tissue shrinkage
  • Dries slowly with no residue
  • Less toxic than xylene
  • Soluble with alcohol, paraffin embedding media, and mounting media
  • Biodegradable
Xylene substitute for histology and cytology is a safer alternative to xylene. Can be used as a solvent and clearing agent, as well as for dissolving paraffin waxes, glues and adhesives. It can be used in all procedures that require xylene.