Seamless Tank Liners, Single-Use, Advanced Scientifics
Supplier: Advanced Scientific, Inc.
B100364-I B100373-I B100363-I B101459-I B101460-I
89127-018CS 4656.43 CAD
89127-018 89127-020 89127-022 89127-024 89127-026
Seamless Tank Liners, Single-Use, Advanced Scientifics
Bags Media Bags
ASI's single-use tank liners are manufactured in an ISO Class 7 environment, packaged clean, irradiated and delivered sterile.
  • Available to improve process for open tank applications
The use of liners eliminates cleaning and turnover concerns associated with reusable stainless tanks. Tank liners offer seamless, streamlined fit, which eliminates pooling areas or 'pleats'.