SP Bel-Art H-B DURAC® Plus™ Pocket Liquid-In-Glass Thermometers, Organic Liquid Fill, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B60770-1800 B60770-1100 B60770-0300 B60770-0400
30617-776EA 39.48 CAD
30617-776 30617-814 30617-818 30617-820 30617-824 30617-826 30617-830
SP Bel-Art H-B DURAC® Plus™ Pocket Liquid-In-Glass Thermometers, Organic Liquid Fill, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
Featuring a case to protect from breakage, these total immersion thermometers are a compact 160mm (6.3”), making them ideal for field studies, waste water treatment and any “on-the-go” applications
  • Mercury free; contain blue organic liquid against lead-free, white back glass
  • Tested and inspected by our USA quality team
  • Traceable to NIST
Case options include Plastic Window - encased in plastic with an open window, Closed Plastic - housed in removable plastic case, and Closed Metal - housed in a removable metal case.
Ordering information: Includes multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.