SP Bel-Art Enviro-Safe® Pocket Liquid-in-Glass Thermometers with Biodegradable Liquid, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B60570-1300 B60570-1400 B60570-1000 B60570-1500
30617-812EA 48.95 CAD
30617-812 30617-816 30617-822 30617-828 30617-834
SP Bel-Art Enviro-Safe® Pocket Liquid-in-Glass Thermometers with Biodegradable Liquid, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
EnviroKleen™ certified thermometers feature green, non-toxic, biodegradable, Enviro-Safe® liquid against lead-free, white back glass
  • Total immersion pocket thermometers are a compact 160mm (6.3”) long
  • Come with a case to protect from breakage
  • Ideal for field studies, waste water treatment and any “on-the-go” applications
  • Traceable to NIST
Should you require an individualized Thermometer Calibration Report, custom calibration services are available.
Certifications: Each item is tested and calibrated in our exclusive triple accredited/registered ISO/More Product Information