Pierce™ Nickel Coated Plates, Thermo Scientific
Supplier: Thermo Scientific
15142 15242 15342 15442
CAPI15142PK 684.01 CAD
CAPI15142 CAPI15242 CAPI15342 CAPI15442
Pierce™ Nickel Coated Plates, Thermo Scientific
Microplates Immunology Microplates and Strips
These Ni2+ chelate-coated plates are ideal for analyzing polyhistidine-tagged fusion proteins by ELISA-based methods.
  • Ni2+ activated surface enables metal-chelate binding of His-tagged proteins
  • Detergents used to lyse cells don't inhibit binding to activated plates as they do with plain polystyrene
  • Binding capacity: ~9 pmol His-tagged protein (27 kDa) per well
Proteins that contain a succession of several histidine residues at the amino or carboxyl terminus have a strong binding affinity for metal
Detergents used to lyse cells do not inhibit binding to Ni-coated plates as they do with plain polystyrene. The detection limit is 1 ng of histidine fusion protein.
These plates provide better binding for more sensitive assays compared to other commercially available nickel-activated plates.
Bacterial lysates containing polyhistidine-tagged fusion proteins can be added directly to the plates without the need for blocking. The clear, white or black plates can be used with colorimetric, chemiluminescent or fluorescent detection methods, respectively.