Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Supplier: Thermo Fisher Scientific
810001 9100CB 900021-WA 900022-WA 900023 900024 910001 911110 911125 900020 910060
14004-272EA 174.44 CAD
14004-272 23198-300 34104-296 34104-298 34104-300 34104-302 34108-041 34175-516 34175-524 34104-294 10781-884
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Electrolytes Electrode Storage Solutions
Cleaning Solutions restore proper electrode performance and prolong the useful life of the electrode.
  • Restore electrode performance
  • Prolong usefulness
  • Unique formulations
Cleaning Solutions are identified by the letters A, B, C, and D. Each letter designates a unique formulation. Cleaning solution A is for removing protein contaminants. Cleaning solution B is for removing bacterial contaminants. Cleaning solution C is for general cleaning. Cleaning solution D is for removing oil and grease contaminants. Bottles include a small beaker to hold the solution and a plastic pipet to remove the old electrode filling solution.