Pierce™ Micro BCA™ Protein Assay Kit and Reagents, Thermo Scientific
Supplier: Thermo Scientific
Pierce
23231 23234 23232 23232 23235 23235 23234 23231
CAPI23234EA 108.59 CAD
PI23234 PI23235 CAPI23234 CAPI23231 PI23231 CAPI23232 PI23232 CAPI23235
Pierce™ Micro BCA™ Protein Assay Kit and Reagents, Thermo Scientific
Assays Protein Assays
The Micro BCA Protein Assay Kit is a specialized version of the popular Pierce BCA Protein Assay for determining the protein concentration of dilute samples.
  • Colorimetric method read at 562 nm
  • Linear working range for BSA is 0.5 to 20 µg/mL
  • Compatible with most ionic and non ionic detergents
  • Less protein variation than dye-binding methods
  • All kit reagents stable at room temperature for two years
  • Working reagent is stable for 24 hours
  • Convenient for microplate or cuvette format
The assay is exceptionally linear and exhibits very low levels of protein-to-protein variability.
This kit is sensitive, colorimetric, stable at room temerature for 24 horus, and compatible with typical concentrations of most ionic and nonionic detergents.