Kjeldahl Tablets, Free of Mercury and Selenium, MilliporeSigma
Supplier: MilliporeSigma
1.15348.0250
1.15348.1000
1.18348.0250
CA1.15348.0250CS
13290
CAD
CA1.15348.0250
CA1.15348.1000
CA1.18348.0250
Kjeldahl Tablets, Free of Mercury and Selenium, MilliporeSigma
Digestion Systems
Kjeldahl Tablets and Reagents
Free of mercury and selenium
For Kjeldahl Analysis and Nitrogen Determination. Titanium(IV) oxide, sodium sulphate, copper(II) sulphate, potassium sulphate.