SP Bel-Art Book Bottles with Spigot, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H11862-0005 H11862-0010
47750-738EA 356.29 CAD
47750-738 47750-740
SP Bel-Art Book Bottles with Spigot, Bel-Art Products, a part of SP
Bottles Carboys
Polypropylene book bottle-style container has a gasketed spigot threaded into the bottle.
  • Polypropylene construction
  • Front is graduated markings in liters
  • Easy to use
It requires only a ¼" turn from closed position to full open and has a 9.5 mm (⅜") Int.Ø.
Note: Spigot has a rubber component that is incompatible with acetone and organic solvents.
opening that extends from the front of the bottle by 58 mm (2.3"). The bottle has a 43 mm Int.Ø neck and features a leakproof seal plug as well as a screw cap.