BugBuster® Master Mix, MilliporeSigma
Supplier: MilliporeSigma
71456-4 71456-3
CA82601-024EA 2030.28 CAD
CA82601-024 CA82601-026
BugBuster® Master Mix, MilliporeSigma
Protein Extraction Protein Extraction Kits
BugBuster® Master Mix combines BugBuster Protein Extraction Reagent with Benzonase® Nuclease and rLysozyme™ Solution in one convenient reagent
BugBuster Master Mix allows for maximum recovery of active soluble protein from both Gram-negative and Gram-positive bacteria. With the Master Mix, there is no need for dilution or separate addition steps. The two available package sizes provide sufficient reagents for protein extraction from 20 g and 100 g cell paste.