n-Hexane ≥99.8%, SupraSolv® for gas chromatography, for chromatography, distilled, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

1.00795.1000 1.00795.2500
CA1.00795.1000CS 2329 CAD
CA1.00795.1000 CA1.00795.2500
n-Hexane ≥99.8%, SupraSolv® for gas chromatography, for chromatography, distilled, Supelco®
n-Hexane

SupraSolv® MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results. This method is used e. g. for the analysis of dioxins and furans (PCDD/PCDF) in food and water samples or for the determination of PAH (polycyclic aromatic hydrocarbons) in food.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: -94.3
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) (sum of hexane isomers plus methylcyclopentane) =98.0% (=99.8%)
Identity(IR) Conforms
Evaporation Residue = 3.0 mg/l
Water = 0.01%
Color =10 Hazen
GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)) = 3ng/ml
Suitability Residue Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR