Ethyl acetate ≥99.8%, SupraSolv® for GC-MS, distilled, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.00789.1000
1.00789.2500
CA1.00789.1000CS
1297
CAD
CA1.00789.1000
CA1.00789.2500
Ethyl acetate ≥99.8%, SupraSolv® for GC-MS, distilled, Supelco®
Ethyl acetate
SupraSolv® MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results. This method is used e. g. for the analysis of dioxins and furans (PCDD/PCDF) in food and water samples or for the determination of PAH (polycyclic aromatic hydrocarbons) in food.
Formula:
CH₃COOC₂H₅ Boiling Pt: 77.1 °C (1013 hPa) Melting Pt: –83 °C Density: 0.902 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009171 CAS Number: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3,II REACH: 01-2119475103-46 Merck Index: 13,03792 |
|
Specification Test Results
Purity (GC) |
|
Identity(IR) | Conforms |
Evaporation Residue |
|
Water |
|
Color |
|
GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)) |
|
Suitability | Residue Analysis |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...