Acetonitrile ≥99.8%, SupraSolv® for GC-MS, distilled, Supelco®
Supplier: MilliporeSigma
|
Danger
|
SupraSolv® MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results. This method is used e. g. for the analysis of dioxins and furans (PCDD/PCDF) in food and water samples or for the determination of PAH (polycyclic aromatic hydrocarbons) in food. Also commonly used in QuEChERS pesticide residue analysis.
Formula:
H₃CCN Boiling Pt: 81.6 °C (1013 hPa) Melting Pt: –45.7 °C Density: 0.782 g/cm³ (20 °C) Flash Pt: 2 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00001878 CAS Number: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
|
Specification Test Results
Purity (GC) |
|
Identity(IR) | Conforms |
Evaporation Residue |
|
Water |
|
Color |
|
GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)) |
|
Suitability | Residue Analysis |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...