Acetone ≥99.8%, SupraSolv® for GC-MS, distilled, Supelco®

Supplier: MilliporeSigma
Danger

1.00658.1000 1.00658.2500
CA1.00658.1000CS 1299 CAD
CA1.00658.1000 CA1.00658.2500
Acetone ≥99.8%, SupraSolv® for GC-MS, distilled, Supelco®
Acetone

SupraSolv® MS is dedicated for use in gas chromatography coupled with mass spectrometric detection. SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results. This method is used e. g. for the analysis of dioxins and furans (PCDD/PCDF) in food and water samples or for the determination of PAH (polycyclic aromatic hydrocarbons) in food.

Formula: CH₃COCH₃
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) =99.8%
Identity(IR) Conforms
Evaporation Residue = 3.0 mg/l
Water = 0.05%
Color =10 Hazen
GC/MSD (retention range n-undecane to n-tetracontane, scanning area 30-600 amu, individual signals (n-tetradecane standard)) = 3ng/ml
Suitability Residue Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR