96-Well PCR Tube Racks
Supplier: Heathrow Scientific
HS23461A HS23461D HS23461B HS23461C
71003-652CS 5833.22 CAD
71003-652 490007-632 76534-768 76534-770
96-Well PCR Tube Racks
Racks Tube Racks
Perfect for pre- or post-PCR sampling.
  • PCR rack is made of polypropylene
  • Accommodates individual tubes or strips
  • Removable hinged lid lies flat
  • Alphanumeric identification for quick location and retrieval of tubes
These sturdy polypropylene racks accommodate individual 0.2 ml tubes.
Ordering information: Assorted color orders include blue, green, orange, purple, and yellow racks.