Foil Rolls
Supplier: Heathrow Scientific
HS23534A HS23536A
89234-756EA 34.9 CAD
89234-756 89234-758
Foil Rolls
Aluminium Foils
Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes, and petri dishes.
  • Create custom-size pieces of foil
  • Two sizes available
These foil rolls are ideal for autoclaving, freezing, incubating, protecting lab ware, lining, sealing, and general storage.