Corning® LSE™ Digital Microplate Shaker, Corning
Supplier: Corning
Corning®
6780-4
89133-526CS 3040.15 CAD
89133-526
Corning® LSE™ Digital Microplate Shaker, Corning
Shakers Microplate Shakers
The Corning LSE Digital Microplate Shaker features digital control of shaking speed and run time
A single control knob is used to set parameters, and values are shown on a large 3-digit display. A sturdy base encloses the motor for quiet, vibration-free operation. The included stainless steel platform accommodates up to 4 microplates (standard or deep well) and the shaker is safe for use in temperature controlled environments from 4 to 65°C. Digital control panel.
Certifications: CE Compliant, CSA Compliant.