Xylene (mixture of isomers) 98,5 ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Dimethylbenzene

97244-700 97244-940
CA71009-212EA 117.19 CAD
CA71009-212 CA71009-220 CA71009-224
Xylene (mixture of isomers) 98,5 ACS
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 25
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Xylene isomers plus Ethylbenzene) 98.5% min.
Ethylbenzene 25% max.
Colour (APHA) 10 max.
Residue after evaporation 0.002% max.
Substances darkened by H2SO4 Passes Test
Sulfur compounds (as S) 0.003% max.
Water (H2O) 0.05% max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR