Sulfuric acid 95.0-98.0% ACS

Supplier: Anachemia

88366-461 88366-543 5300-6G250C 88366-460 88366-461 88366-463
CA71008-946CS 790.25 CAD
80722-392 CA71008-946 CA11024-170 71008-946 CA71008-950 CA71008-948
Sulfuric acid 95.0-98.0% ACS
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II
Merck Index: 14,08974

Order Now

Specification Test Results

Assay (H₂SO₄) 95.0 - 98.0%
Color (APHA) Max. 10
Residue after Ignition Max. 5 ppm
Chloride (Cl) Max. 0.2 ppm
Nitrate (NO₃) Max. 0.5 ppm
Ammonium (NH₄) Max. 2 ppm
Substance Reducing KMnO₄ (as SO₂) Max. 2 ppm
Arsenic (As) Max. 0.01 ppm
Heavy Metals (as Pb) Max. 1 ppm
Iron (Fe) Max. 0.2 ppm
Mercury (Hg) Max. 5 ppb
Appearance Free from Suspended or Insoluble Matter
Identification To pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR