Sodium nitrate ≥99.0%, crystals ACS
Supplier: VWR
|
Warning
|
83720-300
CA71008-746EA
246.91
CAD
CA71008-746
Sodium nitrate ≥99.0%, crystals ACS
Sodium nitrate
Formula:
NaNO₃ Boiling Pt: 380 °C (1013 hPa) Melting Pt: 308 °C Density: 2.26 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011119 CAS Number: 7631-99-4 EINECS: 231-554-3 UN: 1498 ADR: 5.1,III REACH: 01-2119488221-41 Merck Index: 13,08720 |
Specification Test Results
Assay (NaNO3) | 99.0% min. |
pH of a 5% Solution at 25°C | 5.5 - 8.3 |
Insoluble Matter | 0.005% max. |
Chloride (Cl) | 0.001% max. |
Iodate (IO3) | 5 ppm max. |
Nitrite (NO2) | 0.001% max. |
Phosphate (PO4) | 5 ppm max. |
Sulfate (SO4) | 0.003% max. |
Calcium (Ca) | 0.005% max. |
Magnesium (Mg) | 0.002% max. |
Heavy metals (as Pb) | 5 ppm max. |
Iron (Fe) | 3 ppm max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...