Sodium hydroxide 49,0 - 51,0% (w/w) in aqueous solution

Supplier: VWR
Danger

Synonyms: Caustic soda

83082-540 83082-700 83082-940
CA71008-652CS 1070.56 CAD
CA71008-652 CA71008-656 CA71008-660
Sodium hydroxide 49,0 - 51,0% (w/w) in aqueous solution
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Assay (NaOH) 49.0 - 51.0% w/w
Sodium carbonate (Na2CO3) 0.3% max.
Chloride (Cl) 0.01% max.
Nitrogen compounds (as N) 5 ppm max.
Phosphate (PO4) 5 ppm max.
Sulfate (SO4) 0.002% max.
Heavy metals (as Ag) 0.001% max.
Iron (Fe) 5 ppm max.
Potassium (K) 0.05% max.
Nickel (Ni) 0.001% max.
Appearance Clear, colourless solution


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR