Sodium hydroxide ≥97.0%, pellets ACS
Supplier: VWR
|
Danger
|
Synonyms:
Caustic soda
83076-300
83076-380
83076-580
83076-760
CA71008-636EA
122.73
CAD
CA71008-636
CA71008-638
CA71008-640
CA71008-644
Sodium hydroxide ≥97.0%, pellets ACS
Sodium hydroxide
Formula:
NaOH Boiling Pt: 1390 °C (1013 hPa) Melting Pt: 323 °C Density: 2.13 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,08627 |
Specification Test Results
Assay (NaOH) | 97.0% min. |
Sodium carbonate (Na2CO3) | 1.0% max. |
Chloride (Cl) | 0.005% max. |
Nitrogen compounds (as N) | 0.001% max. |
Phosphate (PO4) | 0.001% max. |
Sulfate (SO4) | 0.003% max. |
Heavy metals (as Ag) | 0.002% max. |
Iron (Fe) | 0.001% max. |
Mercury (Hg) | 0.1 ppm max. |
Nickel (Ni) | 0.001% max. |
Calcium (Ca) | 0.005% max. |
Magnesium (Mg) | 0.002% max. |
Potassium (K) | 0.02% max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...