Sodium chloride ≥99.0%, crystals ACS

Supplier: VWR

Synonyms: HaliteSea saltNaClSaline

ANAC81708-820 ANAC81708-580
CA71008-568EA 699.5 CAD
CA71008-568 CA71008-572
Sodium chloride ≥99.0%, crystals ACS
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now

Specification Test Results

Assay (NaCl) 99.0% min.
pH of a 5% Solution at 25°C 5.0 - 9.0
Insoluble Matter 0.005% max.
Iodide (I) 0.002% max.
Bromide (Br) 0.01% max.
Chlorate and nitrate (as NO3) 0.003% max.
Phosphate (PO4) 5 ppm max.
Sulfate (SO4) 0.004% max.
Barium(Ba) Passes Test
Heavy metals ( as Pb) 5 ppm max.
Iron (Fe) 2 ppm max.
Calcium (Ca) 0.002% max.
Magnesium (Mg) 0.001% max.
Potassium (K) 0.005% max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR