Silver sulfate, powder ACS

Supplier: VWR

Synonyms: Disilver sulphateSilver(I) sulfateDisilver sulfate

ANAC79764-300
CA71008-460EA 10326.77 CAD
CA71008-460
Silver sulfate, powder ACS
Silver sulphate
Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: 1085 °C (1013 hPa)
Melting Pt: 655 °C
Density: 5.45 g/cm³ (20 °C)
MDL Number: MFCD00003407
CAS Number: 10294-26-5
Merck Index: 13,08603
https://ca.vwr.com/stibo/web/std.lang.all/94/57/2299457.jpg

Order Now

Specification Test Results

Assay (Ag2SO4) 98.0% min.
Insoluble Matter and AgCl 0.02% max.
Nitrate (NO3) 0.001% max.
Substances not precipitated by HCl 0.03% max.
Iron (Fe) 0.001% max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR