Silver nitrate 0.1 N in aqueous solution volumetric solution, traceable to NIST Standard Reference Material (SRM)

Supplier: VWR
Warning

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

ANAC79692-540 ANAC79692-700
80721-706CS 1442.9 CAD
80721-706 80721-708 80721-710 80721-712
Silver nitrate 0.1 N in aqueous solution volumetric solution, traceable to NIST Standard Reference Material (SRM)
Silver nitrate
Formula: AgNO₃
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance A clear, colourless solution
Normality 0.095 - 0.105 N
Traceable to NIST standards


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR