2-Propanol ≥99.5% ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

76406-543 76406-690 76406-700 76406-540
CA71008-340CS 826.39 CAD
CA71008-340 CA71008-342 CA71008-346 CA71008-348 CA71008-354 CA11020-580 CA11027-466
2-Propanol ≥99.5% ACS
2-Propanol
Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (CH3CHOHCH3) 99.5% min.
Carbonyl compounds:
Propionaldehyde (by GC) 0.002% max.
Acetone (by GC) 0.002% max.
Color (APHA) 10 max.
Residue after evaporation 0.001% max.
Solubility in water Passes Test
Water (H2O) 0.2% max.
Titrable Acid or Base 0.0001 meq/g max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR