Phenolphthalein, powder ACS

Supplier: Avantor

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

ANAC68218-300
CA71007-972EA 375.5 CAD
CA71007-972
Phenolphthalein, powder ACS
Phenolphthalein
Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Melting Pt: 258…262 °C
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00005913
CAS Number: 77-09-8
Merck Index: 14,07243
https://ca.vwr.com/stibo/web/std.lang.all/72/01/2297201.jpg

Order Now

Specification Test Results

Clarity of alcohol solution Passes Test
Visual transition interval pH 8.0(colourless) - 10(red)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR